Learn More
MSC SPI Digital Force Gage, 1000 N, 0.1 N, +/-0.2% FS
Catalog No. 5000535490
Encompass
This digital force gage measures compression and tension, providing precise force measurement for diverse applications. It offers high capacity, excellent resolution, and accurate readings, ideal for professional use in laboratories and industrial settings.
  • Measures compression and tension
  • Selectable units: N, kgf, lbf, gf, ozf, mN
  • Reverse display setting
  • Overload protection mechanism
  • Touch key operation
  • Mechanical overload protection for compression
  • Supports curve or digital display modes
  • Automatic power off
  • Durable metal case
  • 1000 data memory
  • Measurement modes: track, peak, preset peak
  • 160x128 dot matrix LCD (2.8 inch) with LED backlight
  • Powered by rechargeable batteries
Catalog No. 50-005-35490 Supplier MSC Supplier No. 85391464
This digital force gage measures compression and tension, providing precise force measurement for diverse applications. It offers high capacity, excellent resolution, and accurate readings, ideal for professional use in laboratories and industrial settings.
  • Measures compression and tension
  • Selectable units: N, kgf, lbf, gf, ozf, mN
  • Reverse display setting
  • Overload protection mechanism
  • Touch key operation
  • Mechanical overload protection for compression
  • Supports curve or digital display modes
  • Automatic power off
  • Durable metal case
  • 1000 data memory
  • Measurement modes: track, peak, preset peak
  • 160x128 dot matrix LCD (2.8 inch) with LED backlight
  • Powered by rechargeable batteries