Learn More
MSC SPI Drill Point Gage, Stainless Steel, 118 Degree Bevel, with Ruler
Catalog No. 5000535871
Encompass
This drill point gage is designed for precise measurement and verification of twist drills, ensuring accuracy in drilling operations. It features a robust stainless steel head and is specifically calibrated for 118-degree bevel angles, a common standard for general-purpose drills.
  • Head made of stainless steel
  • Bevel angle of 118 degrees
  • Includes a ruler for measurements
  • Ruler measures 15 mm in width and 20 mm in length
  • Graduation located on the front of the ruler
  • Features a smooth surface for ease of use
  • Suitable for use with twist drills that have a 118-degree angle
Catalog No. 50-005-35871 Supplier MSC Supplier No. 83778456
This drill point gage is designed for precise measurement and verification of twist drills, ensuring accuracy in drilling operations. It features a robust stainless steel head and is specifically calibrated for 118-degree bevel angles, a common standard for general-purpose drills.
  • Head made of stainless steel
  • Bevel angle of 118 degrees
  • Includes a ruler for measurements
  • Ruler measures 15 mm in width and 20 mm in length
  • Graduation located on the front of the ruler
  • Features a smooth surface for ease of use
  • Suitable for use with twist drills that have a 118-degree angle