Learn More
MSC SPI Drop Indicator Ball Needle Contact Point, #4-48 Thread, 0.7500 in Point, Chrome Alloy Steel
Catalog No. 5000512571
Encompass
This product is an SPI Drop Indicator Ball Needle Contact Point designed for use with Dial Indicators. It features a #4-48 thread, a 0.7500 inch point length, and is constructed from durable Chrome Alloy Steel. The ball needle contact point style has a ball diameter of 0.1875 inches (3/16 inch) and meets AGD Group 1 industry designation.
  • Absolute encoder ensures all settings
  • Switchable resolutions on models noted below
  • Function lock
  • Designed for use with dial indicators
Catalog No. 50-005-12571 Supplier MSC Supplier No. 35915420
This product is an SPI Drop Indicator Ball Needle Contact Point designed for use with Dial Indicators. It features a #4-48 thread, a 0.7500 inch point length, and is constructed from durable Chrome Alloy Steel. The ball needle contact point style has a ball diameter of 0.1875 inches (3/16 inch) and meets AGD Group 1 industry designation.
  • Absolute encoder ensures all settings
  • Switchable resolutions on models noted below
  • Function lock
  • Designed for use with dial indicators