Learn More
MSC SPI Drop Indicator Ball Needle Contact Point, Chrome Alloy Steel, #4-48 Thread, 0.7500 in Length
Catalog No. 5000527740
Encompass
The SPI Drop Indicator Ball Needle Contact Points are designed for precise measurements when used with Dial Indicators. Constructed from durable chrome alloy steel, these contact points provide reliable performance for various applications.
  • Ball needle contact point style
  • Manufactured from chrome alloy steel
  • Features a contact point length of 0.7500 in
  • Equipped with a ball diameter of 0.1250 in
  • Thread size is #4-48
  • Industry designation is AGD Group 1
Catalog No. 50-005-27740 Supplier MSC Supplier No. 35915446
The SPI Drop Indicator Ball Needle Contact Points are designed for precise measurements when used with Dial Indicators. Constructed from durable chrome alloy steel, these contact points provide reliable performance for various applications.
  • Ball needle contact point style
  • Manufactured from chrome alloy steel
  • Features a contact point length of 0.7500 in
  • Equipped with a ball diameter of 0.1250 in
  • Thread size is #4-48
  • Industry designation is AGD Group 1