Learn More
MSC SPI Drop Indicator Contact Point, Ball Needle, #4-48 Thread, 0.7500 in, Chrome Alloy Steel
Catalog No. 5000528157
Encompass
These drop indicator ball needle contact points are designed for precise measurements with dial indicators. They are constructed from durable chrome alloy steel, featuring a specific thread size and point length for accurate readings.
  • Ball needle contact point style
  • Made from chrome alloy steel
  • Point length of 0.7500 in
  • #4-48 thread size
  • AGD Group 1 industry designation
  • Designed for use with dial indicators
Catalog No. 50-005-28157 Supplier MSC Supplier No. 35915461
These drop indicator ball needle contact points are designed for precise measurements with dial indicators. They are constructed from durable chrome alloy steel, featuring a specific thread size and point length for accurate readings.
  • Ball needle contact point style
  • Made from chrome alloy steel
  • Point length of 0.7500 in
  • #4-48 thread size
  • AGD Group 1 industry designation
  • Designed for use with dial indicators