Learn More
MSC SPI Drop Indicator Contact Point, Solid Carbide, #4-48 Thread, 0.5000 in
Catalog No. 5000512070
Encompass
This cupped ball contact point is designed for use with dial indicators. It features a solid carbide construction and a #4-48 thread, with a point length of 0.5000 inches, making it suitable for precise measurement applications.
  • Cupped ball contact style
  • Solid carbide point material
  • 0.5000 inch point length
  • #4-48 thread size
  • AGD Group 1 industry designation
  • Designed for use with dial indicators
Catalog No. 50-005-12070 Supplier MSC Supplier No. 35915198
This cupped ball contact point is designed for use with dial indicators. It features a solid carbide construction and a #4-48 thread, with a point length of 0.5000 inches, making it suitable for precise measurement applications.
  • Cupped ball contact style
  • Solid carbide point material
  • 0.5000 inch point length
  • #4-48 thread size
  • AGD Group 1 industry designation
  • Designed for use with dial indicators