Learn More
MSC SPI Drop Indicator Radius Contact Point, #4-48 Thread, 1.0000 in Point Length, Steel
Catalog No. 5000528651
Encompass
These contact points are designed for precision measurements and are compatible with dial indicators. They feature a radius style and are constructed from durable steel, offering reliable performance for various applications.
  • Designed for use with dial indicators.
  • Radius style contact point.
  • Constructed from steel.
  • #4-48 thread size.
  • 1.0000 inch point length.
  • 3/16 inch diameter.
Catalog No. 50-005-28651 Supplier MSC Supplier No. 35915859
These contact points are designed for precision measurements and are compatible with dial indicators. They feature a radius style and are constructed from durable steel, offering reliable performance for various applications.
  • Designed for use with dial indicators.
  • Radius style contact point.
  • Constructed from steel.
  • #4-48 thread size.
  • 1.0000 inch point length.
  • 3/16 inch diameter.