Learn More
MSC SPI Drop Indicator Radius Contact Point, Steel, #4-48 Thread, 0.2500 in Length
Catalog No. 5000512321
Encompass
This drop indicator radius contact point is designed for precision measurements with dial indicators. Constructed from durable steel, it features a #4-48 thread and a 0.2500 in (1/4 in) point length, meeting AGD Group 1 industry standards for reliable performance.
  • Steel point material
  • #4-48 thread size
  • 0.2500 in (1/4 in) contact point length
  • 3/16 in diameter
  • Designed for use with dial indicators
  • Meets AGD Group 1 industry designation
Catalog No. 50-005-12321 Supplier MSC Supplier No. 35915842
This drop indicator radius contact point is designed for precision measurements with dial indicators. Constructed from durable steel, it features a #4-48 thread and a 0.2500 in (1/4 in) point length, meeting AGD Group 1 industry standards for reliable performance.
  • Steel point material
  • #4-48 thread size
  • 0.2500 in (1/4 in) contact point length
  • 3/16 in diameter
  • Designed for use with dial indicators
  • Meets AGD Group 1 industry designation