Learn More
MSC SPI Electronic Bore Gage, 1 to 1.2 in, 0.000160 in Accuracy, 0.0001 in Resolution
Catalog No. 5000533676
Encompass
SPI Electronic Bore Gages are designed for accurate measurement of internal diameters. This model features self-centering 3 contact points and durable carbide tipped contact points, making it suitable for various laboratory and industrial applications. It operates with single-handed viewing and comes with a calibrated setting ring and an electronic indicator for immediate use.
  • Self-centering 3 contact points for accurate measurement of internal diameters.
  • Single-handed operation, with right and left hand viewing.
  • Highly durable carbide tipped contact points.
  • Supplied with calibrated setting ring.
  • Electronic indicator included.
  • Short form calibration certificate included.
  • Furnished in a fitted case.
  • Two year limited warranty.
  • Includes batteries (CR2032).
Catalog No. 50-005-33676 Supplier MSC Supplier No. 37882800
SPI Electronic Bore Gages are designed for accurate measurement of internal diameters. This model features self-centering 3 contact points and durable carbide tipped contact points, making it suitable for various laboratory and industrial applications. It operates with single-handed viewing and comes with a calibrated setting ring and an electronic indicator for immediate use.
  • Self-centering 3 contact points for accurate measurement of internal diameters.
  • Single-handed operation, with right and left hand viewing.
  • Highly durable carbide tipped contact points.
  • Supplied with calibrated setting ring.
  • Electronic indicator included.
  • Short form calibration certificate included.
  • Furnished in a fitted case.
  • Two year limited warranty.
  • Includes batteries (CR2032).