Learn More
MSC SPI Electronic Bore Gage, 2-2-1/2 in, 0.0002 in Accuracy, Pistol Grip
Catalog No. 5000528383
Encompass
This electronic bore gage delivers precise internal diameter measurements, featuring a 2-2-1/2 inch measuring range, 0.0002 inch accuracy, and 0.0001 inch resolution. It comes equipped with a pistol grip, 3-point contact heads, an electronic indicator, and a calibrated setting ring for ease of use and accuracy.
  • Self-centering 3 contact points for accurate measurement of internal diameters
  • Single-handed operation with right and left hand viewing
  • Highly durable carbide tipped contact points
  • Supplied with a calibrated setting ring
  • Electronic indicator included
  • Short form calibration certificate included
  • Furnished in a fitted case
  • Automatic shut-off
  • Batteries included
  • Accessories included
Catalog No. 50-005-28383 Supplier MSC Supplier No. 37882768
This electronic bore gage delivers precise internal diameter measurements, featuring a 2-2-1/2 inch measuring range, 0.0002 inch accuracy, and 0.0001 inch resolution. It comes equipped with a pistol grip, 3-point contact heads, an electronic indicator, and a calibrated setting ring for ease of use and accuracy.
  • Self-centering 3 contact points for accurate measurement of internal diameters
  • Single-handed operation with right and left hand viewing
  • Highly durable carbide tipped contact points
  • Supplied with a calibrated setting ring
  • Electronic indicator included
  • Short form calibration certificate included
  • Furnished in a fitted case
  • Automatic shut-off
  • Batteries included
  • Accessories included