Learn More
MSC SPI Electronic Bore Gage, 2 to 6 in Measuring Range, 0.00005 in Resolution
Catalog No. 5000529875
Encompass
This electronic bore gage is designed for close tolerance inspection of bores, measuring size, roundness, ovality, and taper. It features a ruggedly protected electronic indicator to minimize shock and offers incremental, absolute, inch, and metric measuring. The gage is furnished in a fitted case and includes essential accessories like a spacer and anvils, making it suitable for various bore inspection needs.
  • Includes an indicator.
  • Features data output.
  • Comes with a 2 year warranty.
  • Equipped with auto shut-off.
  • Furnished in a fitted case.
  • Offers incremental, absolute, inch, and metric measuring.
  • Includes a spacer and anvils.
  • Requires a CR2032 battery, which is included.
  • Comes with 6 anvils.
  • Is an 11-piece set.
  • Electronic indicator is fully protected by a rugged cover.
  • Supports 2 point measurement.
Catalog No. 50-005-29875 Supplier MSC Supplier No. 02038818
This electronic bore gage is designed for close tolerance inspection of bores, measuring size, roundness, ovality, and taper. It features a ruggedly protected electronic indicator to minimize shock and offers incremental, absolute, inch, and metric measuring. The gage is furnished in a fitted case and includes essential accessories like a spacer and anvils, making it suitable for various bore inspection needs.
  • Includes an indicator.
  • Features data output.
  • Comes with a 2 year warranty.
  • Equipped with auto shut-off.
  • Furnished in a fitted case.
  • Offers incremental, absolute, inch, and metric measuring.
  • Includes a spacer and anvils.
  • Requires a CR2032 battery, which is included.
  • Comes with 6 anvils.
  • Is an 11-piece set.
  • Electronic indicator is fully protected by a rugged cover.
  • Supports 2 point measurement.