Learn More
MSC SPI Electronic Caliper, 0 to 6 in, 0.0005 in Resolution, Stainless Steel
Catalog No. 5000528078
Encompass
This electronic caliper offers precise measurements for a variety of applications. Constructed from hardened stainless steel, it features a measurement range of 0 to 6 inches (0 to 150 mm) with a resolution of 0.0005 inches (0.01 mm). It is designed for durability and accuracy in professional environments.
  • Caliper and jaws constructed from stainless steel.
  • Offers inside, outside, step, and depth measurement capabilities.
  • Equipped with a thumb wheel for smooth jaw adjustment.
  • Includes a depth measurement rod for versatile use.
  • Comes with a protective case for storage and transport.
  • Powered by an LR44 battery.
  • IP54 rated for protection against dust and splashing water.
Catalog No. 50-005-28078 Supplier MSC Supplier No. 45472123
This electronic caliper offers precise measurements for a variety of applications. Constructed from hardened stainless steel, it features a measurement range of 0 to 6 inches (0 to 150 mm) with a resolution of 0.0005 inches (0.01 mm). It is designed for durability and accuracy in professional environments.
  • Caliper and jaws constructed from stainless steel.
  • Offers inside, outside, step, and depth measurement capabilities.
  • Equipped with a thumb wheel for smooth jaw adjustment.
  • Includes a depth measurement rod for versatile use.
  • Comes with a protective case for storage and transport.
  • Powered by an LR44 battery.
  • IP54 rated for protection against dust and splashing water.