Learn More
MSC SPI Electronic Caliper, 1000 mm, 0.01 mm Resolution, Fine Adjustment
Catalog No. 5000527355
Encompass
This electronic caliper provides precise measurements up to 1000 mm (40 in), offering a resolution of 0.01 mm (0.0005 in). It incorporates a fine adjustment mechanism for accurate slider positioning and an absolute measuring system, eliminating the need to reset zero upon activation. Made from durable stainless steel, it is designed for demanding laboratory and industrial applications.
  • Absolute encoder eliminates zero setting on power up.
  • SPC/USB data output.
  • Fine adjustment for precise slider movement.
  • Hardened stainless steel construction.
  • Inch/metric conversion.
  • Measures up to 1000 mm (40 in) with 0.01 mm (0.0005 in) resolution.
  • IP54 rated.
  • Furnished in fitted case.
  • NIST traceability certificate included.
  • Supports outside and inside measurements.
Catalog No. 50-005-27355 Supplier MSC Supplier No. 69548279
This electronic caliper provides precise measurements up to 1000 mm (40 in), offering a resolution of 0.01 mm (0.0005 in). It incorporates a fine adjustment mechanism for accurate slider positioning and an absolute measuring system, eliminating the need to reset zero upon activation. Made from durable stainless steel, it is designed for demanding laboratory and industrial applications.
  • Absolute encoder eliminates zero setting on power up.
  • SPC/USB data output.
  • Fine adjustment for precise slider movement.
  • Hardened stainless steel construction.
  • Inch/metric conversion.
  • Measures up to 1000 mm (40 in) with 0.01 mm (0.0005 in) resolution.
  • IP54 rated.
  • Furnished in fitted case.
  • NIST traceability certificate included.
  • Supports outside and inside measurements.