Learn More
MSC SPI Electronic Caliper, 200 mm, 0.01 mm Resolution, Stainless Steel
Catalog No. 5000512847
Encompass
This electronic caliper is constructed from hardened stainless steel and is designed for precise inside measurements. It comes calibrated and includes NIST traceability, ensuring reliable and accurate results for various applications.
  • Measures up to 200 mm (8 inches)
  • Provides a resolution of 0.01 mm (0.0005 inches)
  • Ensures accuracy within 0.03 mm (0.0015 inches)
  • Features an IP54 rating for protection against dust and splashes
  • Made from durable hardened stainless steel
  • Includes NIST traceability certification for calibration
Catalog No. 50-005-12847 Supplier MSC Supplier No. 96256383
This electronic caliper is constructed from hardened stainless steel and is designed for precise inside measurements. It comes calibrated and includes NIST traceability, ensuring reliable and accurate results for various applications.
  • Measures up to 200 mm (8 inches)
  • Provides a resolution of 0.01 mm (0.0005 inches)
  • Ensures accuracy within 0.03 mm (0.0015 inches)
  • Features an IP54 rating for protection against dust and splashes
  • Made from durable hardened stainless steel
  • Includes NIST traceability certification for calibration