Learn More
MSC SPI Electronic Caliper, 450 mm, 0.01 mm Resolution, Stainless Steel
Catalog No. 5000531234
Encompass
This electronic caliper provides precise measurements with fine adjustment for accurate slider movement. It features nib-shaped jaws for both inside (ID) and outside (OD) measurements, offers inch/metric conversion, and allows zero setting at any position. Built from durable stainless steel, it is suitable for various laboratory and industrial applications.
  • Fine adjustment for precise slider movement.
  • Nib shaped jaws for inside and outside diameter measurements.
  • Carrying upper jaws for internal measurements.
  • Inch/metric conversion with zero setting at any position.
  • Data output enabled via SPC cable, wireless transmitter, or Micro USB.
  • IP54 rated.
  • Constructed from durable stainless steel.
  • Includes a plastic box.
  • Meets ASME B89.1.14 standards.
Catalog No. 50-005-31234 Supplier MSC Supplier No. 83778449
This electronic caliper provides precise measurements with fine adjustment for accurate slider movement. It features nib-shaped jaws for both inside (ID) and outside (OD) measurements, offers inch/metric conversion, and allows zero setting at any position. Built from durable stainless steel, it is suitable for various laboratory and industrial applications.
  • Fine adjustment for precise slider movement.
  • Nib shaped jaws for inside and outside diameter measurements.
  • Carrying upper jaws for internal measurements.
  • Inch/metric conversion with zero setting at any position.
  • Data output enabled via SPC cable, wireless transmitter, or Micro USB.
  • IP54 rated.
  • Constructed from durable stainless steel.
  • Includes a plastic box.
  • Meets ASME B89.1.14 standards.