Learn More
MSC SPI Electronic Depth Gage, 0-150 mm, 0.02 mm Accuracy, ABS Case
Catalog No. 5000527980
Encompass
This electronic depth gage offers precise measurements with a 0-150 mm range, 0.02 mm accuracy, and 0.01 mm resolution. It is designed for laboratory and industrial use, featuring automatic on/off to conserve battery life and inch/metric conversion for versatility. The gage includes NIST Traceability Certification and is supplied in a durable fitted case for protection.
  • 0.02 mm accuracy
  • 0.01 mm resolution
  • 4 in base length
  • NIST traceability certification included
  • SPC/USB data output
  • Automatic shut-off
  • Linear scale embedded
  • Inch/metric conversion
  • Absolute and incremental measuring modes
  • Available with or without hook
  • Measurements can be taken on either side of the hook
  • Furnished in fitted case
  • Two year limited warranty
Catalog No. 50-005-27980 Supplier MSC Supplier No. 69547578
This electronic depth gage offers precise measurements with a 0-150 mm range, 0.02 mm accuracy, and 0.01 mm resolution. It is designed for laboratory and industrial use, featuring automatic on/off to conserve battery life and inch/metric conversion for versatility. The gage includes NIST Traceability Certification and is supplied in a durable fitted case for protection.
  • 0.02 mm accuracy
  • 0.01 mm resolution
  • 4 in base length
  • NIST traceability certification included
  • SPC/USB data output
  • Automatic shut-off
  • Linear scale embedded
  • Inch/metric conversion
  • Absolute and incremental measuring modes
  • Available with or without hook
  • Measurements can be taken on either side of the hook
  • Furnished in fitted case
  • Two year limited warranty