Learn More
MSC SPI Electronic Depth Gage, 0 to 8 in, 0.0015 in Accuracy
Catalog No. 5000528283
Encompass
This electronic depth gage provides precise measurements for various applications. It offers a measurement range of 0 to 8 in with an accuracy of 0.0015 in and a resolution of 0.0005 in. Constructed from stainless steel, it features a 4 in base length for stable readings.
  • Accuracy of 0.0015 in
  • Resolution of 0.0005 in
  • 4 in base length for stable measurements
  • Durable stainless steel construction
  • SPC/USB data output capability
  • CR2032 battery included
  • Includes hook attachment
  • IP54 water resistance rating
Catalog No. 50-005-28283 Supplier MSC Supplier No. 35650514
This electronic depth gage provides precise measurements for various applications. It offers a measurement range of 0 to 8 in with an accuracy of 0.0015 in and a resolution of 0.0005 in. Constructed from stainless steel, it features a 4 in base length for stable readings.
  • Accuracy of 0.0015 in
  • Resolution of 0.0005 in
  • 4 in base length for stable measurements
  • Durable stainless steel construction
  • SPC/USB data output capability
  • CR2032 battery included
  • Includes hook attachment
  • IP54 water resistance rating