Learn More
MSC SPI Electronic Outside Micrometer, IP65, 2-3 in / 50-75 mm, Absolute
Catalog No. 5000527183
Encompass
This electronic outside micrometer is designed for precise measurements in the range of 2-3 in / 50-75 mm. It offers a high resolution of .00005 in / .001 mm and an accuracy of +/- 0.0002 in / 0.003 mm.
  • IP65 level protection.
  • Carbide measuring faces.
  • Spindle diameter: 0.250 in.
  • Standards included for all sizes above 0-1 in / 25 mm range.
  • Painted frame with heat shield.
  • Incremental/absolute and inch/metric measuring.
  • Ratchet thimble.
  • Spindle lock.
  • SPC/USB data output.
  • Automatic shut-off.
  • Battery: 357/303 included.
  • Short form certificate of calibration included.
  • Furnished in fitted case.
  • Two year warranty.
Catalog No. 50-005-27183 Supplier MSC Supplier No. 49298946
This electronic outside micrometer is designed for precise measurements in the range of 2-3 in / 50-75 mm. It offers a high resolution of .00005 in / .001 mm and an accuracy of +/- 0.0002 in / 0.003 mm.
  • IP65 level protection.
  • Carbide measuring faces.
  • Spindle diameter: 0.250 in.
  • Standards included for all sizes above 0-1 in / 25 mm range.
  • Painted frame with heat shield.
  • Incremental/absolute and inch/metric measuring.
  • Ratchet thimble.
  • Spindle lock.
  • SPC/USB data output.
  • Automatic shut-off.
  • Battery: 357/303 included.
  • Short form certificate of calibration included.
  • Furnished in fitted case.
  • Two year warranty.