Learn More
MSC SPI Electronic V-Anvil Micrometer, 0.60 in Range, ±.0001 in Accuracy
Catalog No. 5000513426
Encompass
This electronic V-anvil micrometer is designed for precise measurements. It features a ratchet thimble for consistent measuring force, a positive lock nut for secure readings, and the ability to output data, making it suitable for various laboratory and industrial applications.
  • Electronic operation for precision
  • Ratchet thimble for consistent measuring force
  • Positive lock nut for secure measurements
  • Ability to output data
  • Carbide anvil material for durability
Catalog No. 50-005-13426 Supplier MSC Supplier No. 13434998
This electronic V-anvil micrometer is designed for precise measurements. It features a ratchet thimble for consistent measuring force, a positive lock nut for secure readings, and the ability to output data, making it suitable for various laboratory and industrial applications.
  • Electronic operation for precision
  • Ratchet thimble for consistent measuring force
  • Positive lock nut for secure measurements
  • Ability to output data
  • Carbide anvil material for durability