Learn More
MSC SPI Feeler Gage, 0.003 in Thick, 12 in Long, Tempered Steel
Catalog No. 5000527534
Encompass
This feeler gage set is designed for precise measurements, featuring a thickness of 0.003 inch and a length of 12 " Each gage is constructed from durable tempered steel, providing accuracy and longevity for various applications. This product is supplied as a 25 piece set.
  • Precise 0.003 inch thickness
  • 12 inch long leaves
  • Tempered steel construction
  • Comes as a 25 piece set
  • Manufactured in India
Catalog No. 50-005-27534 Supplier MSC Supplier No. 62704879
This feeler gage set is designed for precise measurements, featuring a thickness of 0.003 inch and a length of 12 " Each gage is constructed from durable tempered steel, providing accuracy and longevity for various applications. This product is supplied as a 25 piece set.
  • Precise 0.003 inch thickness
  • 12 inch long leaves
  • Tempered steel construction
  • Comes as a 25 piece set
  • Manufactured in India