Learn More
MSC SPI Feeler Gage, 0.023 in Thick, 0.5 in Wide, 12 in Long, High-Carbon Steel
Catalog No. 5000528959
Encompass
This feeler gage is designed with parallel leaves and is constructed from durable high-carbon steel, ensuring precise measurements and long-lasting performance.
  • Precise thickness of 0.023 "
  • Width of 0.5 "
  • Length of 12 inches.
  • Made from high-carbon steel for durability.
  • Features a parallel leaf style for consistent measurement.
  • Manufactured in Germany.
Catalog No. 50-005-28959 Supplier MSC Supplier No. 85668689
This feeler gage is designed with parallel leaves and is constructed from durable high-carbon steel, ensuring precise measurements and long-lasting performance.
  • Precise thickness of 0.023 "
  • Width of 0.5 "
  • Length of 12 inches.
  • Made from high-carbon steel for durability.
  • Features a parallel leaf style for consistent measurement.
  • Manufactured in Germany.