Learn More
MSC SPI Feeler Gage Stock Roll, 0.90 mm, 1/2 in, 25 ft
Catalog No. 5000535644
Encompass
This feeler gage stock roll is designed for gaging clearance, narrow slots, and gear play. It can also be utilized for shimming in fixture and die work, providing versatility in various applications.
  • Used for gaging clearance, narrow slots, and gear play.
  • Can be used for shimming in fixture and die work.
  • Marked every 6 inches with a line and its thickness.
  • Hardness is RC 48-52.
  • Dimensions are 25 ft long and 1/2 in wide.
  • Supplied in a cardboard box.
Catalog No. 50-005-35644 Supplier MSC Supplier No. 85669422
This feeler gage stock roll is designed for gaging clearance, narrow slots, and gear play. It can also be utilized for shimming in fixture and die work, providing versatility in various applications.
  • Used for gaging clearance, narrow slots, and gear play.
  • Can be used for shimming in fixture and die work.
  • Marked every 6 inches with a line and its thickness.
  • Hardness is RC 48-52.
  • Dimensions are 25 ft long and 1/2 in wide.
  • Supplied in a cardboard box.