Learn More
MSC SPI Feeler Stock Roll, 0.016 in Thick, 1/2 in Wide, High-Carbon Steel
Catalog No. 5000530209
Encompass
This feeler stock roll provides precise measurements for various applications, designed for checking gaps and clearances in industrial and mechanical settings.
  • Provides a thickness of 0.016 "
  • Features a width of 1/2 "
  • Offers an overall length of 25 feet.
  • Constructed from high-carbon steel for durability.
  • Comes with a hassle-free guarantee for exchange or refund within 30 days.
  • Manufactured from first-quality materials from reputable suppliers.
Catalog No. 50-005-30209 Supplier MSC Supplier No. 85669539
This feeler stock roll provides precise measurements for various applications, designed for checking gaps and clearances in industrial and mechanical settings.
  • Provides a thickness of 0.016 "
  • Features a width of 1/2 "
  • Offers an overall length of 25 feet.
  • Constructed from high-carbon steel for durability.
  • Comes with a hassle-free guarantee for exchange or refund within 30 days.
  • Manufactured from first-quality materials from reputable suppliers.