Learn More
MSC SPI Feeler Gage Stock Roll, 0.0400 in., High-Carbon Steel, 25 ft., 1 ea.
Catalog No. 5000514562
Encompass
These feeler gage stock rolls are designed for precise gaging of clearance, narrow slots, and gear play, as well as for shimming in fixture and die work. Constructed from high-carbon steel, these rolls provide durability for rigorous industrial applications.
  • Type C 1095 high-carbon steel construction
  • Hardness rating of 48-52 RC
  • Measurements of 25 ft. long by 1/2 in. wide
  • Thickness of 0.0400 in. with a tolerance of 0.0002 in.
  • Marked every 6 in. with a line and thickness for ease of use
  • Supplied in a cardboard box
Catalog No. 50-005-14562 Supplier MSC Supplier No. 85669778
Add to Cart
Add to Cart
These feeler gage stock rolls are designed for precise gaging of clearance, narrow slots, and gear play, as well as for shimming in fixture and die work. Constructed from high-carbon steel, these rolls provide durability for rigorous industrial applications.
  • Type C 1095 high-carbon steel construction
  • Hardness rating of 48-52 RC
  • Measurements of 25 ft. long by 1/2 in. wide
  • Thickness of 0.0400 in. with a tolerance of 0.0002 in.
  • Marked every 6 in. with a line and thickness for ease of use
  • Supplied in a cardboard box