Learn More
MSC SPI Feeler Stock Roll, High Carbon Steel, 0.001 in Thick
Catalog No. 5000528954
Encompass
This feeler stock roll is made from high-carbon steel and is designed for precision measurements. It provides reliable and accurate thickness for various applications.
  • Made from high carbon steel
  • Provides a thickness of 0.001 inch
  • Measures 1/2 inch in width
  • Comes in a 25 foot roll
Catalog No. 50-005-28954 Supplier MSC Supplier No. 85669448
This feeler stock roll is made from high-carbon steel and is designed for precision measurements. It provides reliable and accurate thickness for various applications.
  • Made from high carbon steel
  • Provides a thickness of 0.001 inch
  • Measures 1/2 inch in width
  • Comes in a 25 foot roll