Learn More
MSC SPI Feeler Stock Roll, High-Carbon Steel, 0.60 mm, 1/2 in
Catalog No. 5000531291
Encompass
This feeler stock roll is made from high-carbon steel, designed for various precision measurement and spacing applications. It provides accurate thickness for gaging and shimming tasks in demanding environments.
  • Made from high-carbon steel for durability and precision
  • Provides 0.60 mm thickness for accurate measurements
  • Features a 1/2 inch width suitable for a range of applications
  • Offers a 10 foot length for ample material
  • Manufactured in Germany, ensuring quality standards
Catalog No. 50-005-31291 Supplier MSC Supplier No. 85669331
This feeler stock roll is made from high-carbon steel, designed for various precision measurement and spacing applications. It provides accurate thickness for gaging and shimming tasks in demanding environments.
  • Made from high-carbon steel for durability and precision
  • Provides 0.60 mm thickness for accurate measurements
  • Features a 1/2 inch width suitable for a range of applications
  • Offers a 10 foot length for ample material
  • Manufactured in Germany, ensuring quality standards