Learn More
MSC SPI Feeler Stock Roll, High-Carbon Steel, 1/2 in, 10 ft
Catalog No. 5000527353
Encompass
This feeler stock roll is made from high-carbon steel, designed for precision measurement and spacing applications. It features a width of 1/2 inch, an overall length of 10 feet, and a thickness of 0.65 mm.
  • High-carbon steel material for durability.
  • Measures 1/2 inch in width.
  • Provides 10 feet of overall length.
  • Offers a precise thickness of 0.65 mm.
  • Suitable for various spacing and gaging tasks.
Catalog No. 50-005-27353 Supplier MSC Supplier No. 85669349
This feeler stock roll is made from high-carbon steel, designed for precision measurement and spacing applications. It features a width of 1/2 inch, an overall length of 10 feet, and a thickness of 0.65 mm.
  • High-carbon steel material for durability.
  • Measures 1/2 inch in width.
  • Provides 10 feet of overall length.
  • Offers a precise thickness of 0.65 mm.
  • Suitable for various spacing and gaging tasks.