Learn More
MSC SPI Feeler Gage, Tempered Steel, 0.03 in. Thick, 0.5 in. Wide, 12 in. Long, 1 ea
Catalog No. 5000514649
Encompass
This tempered steel feeler gage is designed for precise dimensional measurement and gap inspection in metalworking and maintenance applications. It provides a reliable, durable solution for verifying tolerances and clearances.
  • Constructed from durable tempered steel
  • Measures 0.03 in. thick by 0.5 in. wide
  • Features a 12 in. overall leaf length
Catalog No. 50-005-14649 Supplier MSC Supplier No. 62704887
Add to Cart
Add to Cart
This tempered steel feeler gage is designed for precise dimensional measurement and gap inspection in metalworking and maintenance applications. It provides a reliable, durable solution for verifying tolerances and clearances.
  • Constructed from durable tempered steel
  • Measures 0.03 in. thick by 0.5 in. wide
  • Features a 12 in. overall leaf length