Learn More
MSC SPI Gage Ball, 13/16 inch Diameter, Chrome Steel, 0.0001 in Tolerance
Catalog No. 5000528088
Encompass
This gage ball is made of chrome steel and has a diameter of 13/16 " It features a tolerance of 0.0001 inch and a hardness of 63 HRC, ensuring high precision for various measurement and calibration applications.
  • Chrome steel construction
  • Precise 0.0001 inch tolerance
  • Grade 25 accuracy
  • Hardened to 63 HRC for durability
  • AFBMA certified for quality assurance
Catalog No. 50-005-28088 Supplier MSC Supplier No. 06462527
This gage ball is made of chrome steel and has a diameter of 13/16 " It features a tolerance of 0.0001 inch and a hardness of 63 HRC, ensuring high precision for various measurement and calibration applications.
  • Chrome steel construction
  • Precise 0.0001 inch tolerance
  • Grade 25 accuracy
  • Hardened to 63 HRC for durability
  • AFBMA certified for quality assurance