Learn More
MSC SPI Gage Ball, 19/32 in Dia, Chrome Steel, 63 HRC
Catalog No. 5000528445
Encompass
This gage ball is manufactured from chrome steel and features a 19/32 inch diameter with a hardness of 63 HRC. It is designed for precise measurement and inspection applications, offering high accuracy and durability.
  • 19/32 inch diameter
  • Chrome steel material
  • 63 HRC hardness
  • 0.000100 inch tolerance
  • Accuracy grade 25
  • AFBMA certified
  • Hardened heat treatment
Catalog No. 50-005-28445 Supplier MSC Supplier No. 06462386
Add to Cart
Add to Cart
This gage ball is manufactured from chrome steel and features a 19/32 inch diameter with a hardness of 63 HRC. It is designed for precise measurement and inspection applications, offering high accuracy and durability.
  • 19/32 inch diameter
  • Chrome steel material
  • 63 HRC hardness
  • 0.000100 inch tolerance
  • Accuracy grade 25
  • AFBMA certified
  • Hardened heat treatment