Learn More
MSC SPI Gage Ball, 7/8 in, Chrome Steel, 63 HRC, Grade 25
Catalog No. 5000529065
Encompass
Gage balls are precision-engineered inspection tools used for accurate measurement and calibration. This particular gage ball features a 7/8 inch diameter, is constructed from durable chrome steel, and is hardened to 63 HRC for exceptional wear resistance. It meets Accuracy Grade 25, ensuring high precision and reliability for critical inspection applications.
  • Precision-engineered for accurate measurement and calibration.
  • Constructed from durable chrome steel for long-lasting performance.
  • Hardened to 63 HRC for exceptional wear resistance.
  • Meets Accuracy Grade 25 for high precision.
  • Certified, ensuring reliability for critical inspection applications.
  • Ideal for various quality control and metrology tasks.
Catalog No. 50-005-29065 Supplier MSC Supplier No. 06462568
Gage balls are precision-engineered inspection tools used for accurate measurement and calibration. This particular gage ball features a 7/8 inch diameter, is constructed from durable chrome steel, and is hardened to 63 HRC for exceptional wear resistance. It meets Accuracy Grade 25, ensuring high precision and reliability for critical inspection applications.
  • Precision-engineered for accurate measurement and calibration.
  • Constructed from durable chrome steel for long-lasting performance.
  • Hardened to 63 HRC for exceptional wear resistance.
  • Meets Accuracy Grade 25 for high precision.
  • Certified, ensuring reliability for critical inspection applications.
  • Ideal for various quality control and metrology tasks.