Learn More
MSC SPI Gage Ball, Chrome Steel, 63 HRC, 3/16 in Diameter, Accuracy Grade 25
Catalog No. 5000529428
Encompass
This gage ball is constructed from chrome steel and heat-treated for a hardness of 63 HRC. It features a 3/16 inch diameter and an accuracy grade of 25, making it suitable for precise measurement applications.
  • Chrome steel construction
  • Hardness of 63 HRC
  • Accuracy grade of 25
  • AFBMA certified
  • 3/16 inch diameter
  • Heat treated and hardened for durability
Catalog No. 50-005-29428 Supplier MSC Supplier No. 06462121
This gage ball is constructed from chrome steel and heat-treated for a hardness of 63 HRC. It features a 3/16 inch diameter and an accuracy grade of 25, making it suitable for precise measurement applications.
  • Chrome steel construction
  • Hardness of 63 HRC
  • Accuracy grade of 25
  • AFBMA certified
  • 3/16 inch diameter
  • Heat treated and hardened for durability