Learn More
MSC SPI Gage Ball, Chrome Steel, 63 HRC, Grade 25, 11/32 in
Catalog No. 5000512435
Encompass
This gage ball is constructed from chrome steel, featuring a hardness of 63 HRC. It boasts an Accuracy Grade of 25 and is AFBMA Certified, with a diameter of 11/32 inches.
  • Chrome steel material
  • 63 HRC hardness
  • Hardened for increased durability
  • Accuracy grade 25
  • AFBMA certified
  • 11/32 in diameter
Catalog No. 50-005-12435 Supplier MSC Supplier No. 06462220
This gage ball is constructed from chrome steel, featuring a hardness of 63 HRC. It boasts an Accuracy Grade of 25 and is AFBMA Certified, with a diameter of 11/32 inches.
  • Chrome steel material
  • 63 HRC hardness
  • Hardened for increased durability
  • Accuracy grade 25
  • AFBMA certified
  • 11/32 in diameter