Learn More
MSC SPI Gage Ball, Chrome Steel, 63 HRC, Grade 25, 5/32 Inch
Catalog No. 5000529267
Encompass
This SPI Gage Ball is crafted from durable Chrome Steel, engineered for precision measurement applications. It features a hardened construction with a 63 HRC hardness, ensuring reliability and longevity. With an Accuracy Grade of 25 and AFBMA certification, this gage ball provides precise and consistent results for various calibration and inspection tasks.
  • Constructed from high-quality chrome steel for durability.
  • Hardened to 63 HRC for exceptional wear resistance.
  • Features an accuracy grade of 25, ensuring high precision.
  • AFBMA certified for reliable performance standards.
  • Provides a tight tolerance of 0.000100 decimal "
  • Ideal for calibration, layout, and machine setup tools.
Catalog No. 50-005-29267 Supplier MSC Supplier No. 06462105
This SPI Gage Ball is crafted from durable Chrome Steel, engineered for precision measurement applications. It features a hardened construction with a 63 HRC hardness, ensuring reliability and longevity. With an Accuracy Grade of 25 and AFBMA certification, this gage ball provides precise and consistent results for various calibration and inspection tasks.
  • Constructed from high-quality chrome steel for durability.
  • Hardened to 63 HRC for exceptional wear resistance.
  • Features an accuracy grade of 25, ensuring high precision.
  • AFBMA certified for reliable performance standards.
  • Provides a tight tolerance of 0.000100 decimal "
  • Ideal for calibration, layout, and machine setup tools.