Learn More
MSC SPI Gage Ball, Chrome Steel, 9/64 in Diameter, 63 HRC, Grade 25
Catalog No. 5000529063
Encompass
This chrome steel gage ball is hardened to 63 HRC, offering high durability and wear resistance. With a 9/64 inch diameter and an accuracy grade of 25, it provides precise measurements for various inspection applications. This product is also AFBMA certified.
  • Manufactured from chrome steel for durability.
  • Hardness of 63 HRC ensures wear resistance.
  • Features a 9/64 inch diameter for precise gaging.
  • Accuracy grade of 25 for high precision applications.
  • AFBMA certified for quality and standard compliance.
  • Heat treated for enhanced material strength.
  • Has a tolerance of 0.000100 "
Catalog No. 50-005-29063 Supplier MSC Supplier No. 06462097
This chrome steel gage ball is hardened to 63 HRC, offering high durability and wear resistance. With a 9/64 inch diameter and an accuracy grade of 25, it provides precise measurements for various inspection applications. This product is also AFBMA certified.
  • Manufactured from chrome steel for durability.
  • Hardness of 63 HRC ensures wear resistance.
  • Features a 9/64 inch diameter for precise gaging.
  • Accuracy grade of 25 for high precision applications.
  • AFBMA certified for quality and standard compliance.
  • Heat treated for enhanced material strength.
  • Has a tolerance of 0.000100 "