Learn More
MSC SPI Gage Block Bevel End Jaws, 4 in, 0.75 in
Catalog No. 5000514184
Encompass
SPI Gage Block Bevel End Jaws are designed for use with rectangular gage blocks. This set includes two jaws with an overall length of 4 inches and a jaw thickness of 0.75 inches, offering precise measurement capabilities. They feature a flatness accuracy of +/- 0.00002 inches per 2 inches.
  • Bevel end jaws for precise measurements.
  • Designed for use with rectangular gage blocks.
  • 0.75 in jaw thickness.
  • Flatness accuracy of +/- 0.00002 in per 2 in.
  • 4 in overall length.
  • Comes as a set of two pieces.
Catalog No. 50-005-14184 Supplier MSC Supplier No. 50967033
SPI Gage Block Bevel End Jaws are designed for use with rectangular gage blocks. This set includes two jaws with an overall length of 4 inches and a jaw thickness of 0.75 inches, offering precise measurement capabilities. They feature a flatness accuracy of +/- 0.00002 inches per 2 inches.
  • Bevel end jaws for precise measurements.
  • Designed for use with rectangular gage blocks.
  • 0.75 in jaw thickness.
  • Flatness accuracy of +/- 0.00002 in per 2 in.
  • 4 in overall length.
  • Comes as a set of two pieces.