Learn More
MSC SPI Gage Block, Rectangle, 0.1001 in, Steel, Grade AS-1
Catalog No. 5000530242
Encompass
This gage block is a precision measuring tool designed for accurate length reference. It is a rectangular steel block with a nominal size of 0.1001 inches and an AS-1 accuracy grade, suitable for various industrial and laboratory applications requiring high precision.
  • Rectangle shape for versatile use
  • AS-1 accuracy grade ensures high precision
  • Made from durable steel material
  • Meets NIST standards for reliability
  • Nominal size of 0.1001 inches
Catalog No. 50-005-30242 Supplier MSC Supplier No. 87876678
This gage block is a precision measuring tool designed for accurate length reference. It is a rectangular steel block with a nominal size of 0.1001 inches and an AS-1 accuracy grade, suitable for various industrial and laboratory applications requiring high precision.
  • Rectangle shape for versatile use
  • AS-1 accuracy grade ensures high precision
  • Made from durable steel material
  • Meets NIST standards for reliability
  • Nominal size of 0.1001 inches