Learn More
MSC SPI Gage Block, Rectangle, 0.2 in, AS-1 Grade, Steel
Catalog No. 5000534502
Encompass
This steel gage block is designed for precise measurement applications. It features a rectangular shape and an AS-1 accuracy grade, making it suitable for calibration and inspection tasks.
  • Includes NIST certification
  • Rectangle shape
  • AS-1 accuracy grade
  • 0.2 in size
  • Steel material
Catalog No. 50-005-34502 Supplier MSC Supplier No. 87877353
This steel gage block is designed for precise measurement applications. It features a rectangular shape and an AS-1 accuracy grade, making it suitable for calibration and inspection tasks.
  • Includes NIST certification
  • Rectangle shape
  • AS-1 accuracy grade
  • 0.2 in size
  • Steel material