Learn More
MSC SPI Gage Block, Rectangle, 0.5 inch, Grade AS-1
Catalog No. 5000534503
Encompass
This individual rectangular steel gage block is precisely crafted to a size of 0.5 inch and meets AS-1 accuracy grade standards. It is designed for demanding measurement applications, ensuring reliability and precision in various laboratory and industrial settings.
  • Each gage block has blended radii and is burr-proofed.
  • Wringing or stacking the block will not cause damage to the measuring surfaces.
  • Certificate of calibration traceable to NIST is included with each gage.
Catalog No. 50-005-34503 Supplier MSC Supplier No. 87877429
This individual rectangular steel gage block is precisely crafted to a size of 0.5 inch and meets AS-1 accuracy grade standards. It is designed for demanding measurement applications, ensuring reliability and precision in various laboratory and industrial settings.
  • Each gage block has blended radii and is burr-proofed.
  • Wringing or stacking the block will not cause damage to the measuring surfaces.
  • Certificate of calibration traceable to NIST is included with each gage.