Learn More
MSC SPI Gage Block, Rectangle, 0.850 inch, Grade 0, Steel
Catalog No. 5000529884
Encompass
This gage block is designed for precision measurements, featuring a rectangular shape and manufactured from steel. It offers an accuracy grade of 0 and includes NIST certification for reliability.
  • Rectangle shape
  • Accuracy grade 0
  • 0.850 inch size
  • Steel material
  • Includes NIST certification
Catalog No. 50-005-29884 Supplier MSC Supplier No. 03267077
This gage block is designed for precision measurements, featuring a rectangular shape and manufactured from steel. It offers an accuracy grade of 0 and includes NIST certification for reliability.
  • Rectangle shape
  • Accuracy grade 0
  • 0.850 inch size
  • Steel material
  • Includes NIST certification