Learn More
MSC SPI Gage Block, Rectangle, Steel, 0.1003 in, Grade AS-1
Catalog No. 5000510905
Encompass
This rectangle steel gage block features a precise 0.10030 inch size and an AS-1 accuracy grade, making it suitable for high-precision calibration and measurement applications. Constructed from durable steel, this gage block meets NIST standards and includes NIST certification, ensuring reliable and traceable measurements.
  • Precise 0.10030 inch size
  • AS-1 accuracy grade
  • Suitable for high-precision calibration and measurement applications
  • Constructed from durable steel
  • Meets NIST standards
  • Includes NIST certification
Catalog No. 50-005-10905 Supplier MSC Supplier No. 87876694
This rectangle steel gage block features a precise 0.10030 inch size and an AS-1 accuracy grade, making it suitable for high-precision calibration and measurement applications. Constructed from durable steel, this gage block meets NIST standards and includes NIST certification, ensuring reliable and traceable measurements.
  • Precise 0.10030 inch size
  • AS-1 accuracy grade
  • Suitable for high-precision calibration and measurement applications
  • Constructed from durable steel
  • Meets NIST standards
  • Includes NIST certification