Learn More
MSC SPI Gage Block, Rectangle, Steel, 0.132 in, Grade 0
Catalog No. 5000513392
Encompass
This gage block is a precision measuring tool, manufactured from steel and designed for high accuracy applications. It features a rectangular shape and measures 0.132 inches with a Grade 0 accuracy, ensuring reliable and consistent measurements.
  • Each gage block has blended radii and is burr-proofed.
  • Wringing or stacking the block will not cause damage to the measuring surfaces.
  • Certificate of calibration traceable to NIST included with each gage.
Catalog No. 50-005-13392 Supplier MSC Supplier No. 03266756
This gage block is a precision measuring tool, manufactured from steel and designed for high accuracy applications. It features a rectangular shape and measures 0.132 inches with a Grade 0 accuracy, ensuring reliable and consistent measurements.
  • Each gage block has blended radii and is burr-proofed.
  • Wringing or stacking the block will not cause damage to the measuring surfaces.
  • Certificate of calibration traceable to NIST included with each gage.