Learn More
MSC SPI Gage Block, Rectangle, Steel, 0.134 in, Grade 0
Catalog No. 5000512420
Encompass
This gage block is designed for precise measurements, featuring a rectangular shape and constructed from durable steel. It has a size of 0.134 inch and an accuracy grade of 0, including NIST certification to ensure high reliability.
  • Rectangle shape for versatile use.
  • Constructed from durable steel.
  • 0.134 inch size for specific measurement applications.
  • Grade 0 accuracy for high precision.
  • Includes NIST certification for traceable standards.
Catalog No. 50-005-12420 Supplier MSC Supplier No. 03266772
This gage block is designed for precise measurements, featuring a rectangular shape and constructed from durable steel. It has a size of 0.134 inch and an accuracy grade of 0, including NIST certification to ensure high reliability.
  • Rectangle shape for versatile use.
  • Constructed from durable steel.
  • 0.134 inch size for specific measurement applications.
  • Grade 0 accuracy for high precision.
  • Includes NIST certification for traceable standards.