Learn More
MSC SPI Gage Block, Rectangle, Steel, 0.137 Inch, Grade 0
Catalog No. 5000529435
Encompass
This rectangle steel gage block is a precision measurement tool. It features a Grade 0 accuracy, making it suitable for applications requiring high precision. The block is made of steel and comes with NIST certification, ensuring its reliability and traceability for critical measurements.
  • Rectangle shape
  • Grade 0 accuracy
  • Made from durable steel
  • 0.137 inch size
  • Includes NIST certification
Catalog No. 50-005-29435 Supplier MSC Supplier No. 03266806
This rectangle steel gage block is a precision measurement tool. It features a Grade 0 accuracy, making it suitable for applications requiring high precision. The block is made of steel and comes with NIST certification, ensuring its reliability and traceability for critical measurements.
  • Rectangle shape
  • Grade 0 accuracy
  • Made from durable steel
  • 0.137 inch size
  • Includes NIST certification