Learn More
MSC SPI Gage Block, Rectangular, Steel, 0.121 Inch, Grade 0
Catalog No. 5000511443
Encompass
This individual rectangular gage block, crafted from steel, offers precise measurements with a size of 0.121 inch and a Grade 0 accuracy. It is engineered for high durability and reliable performance in diverse measurement applications.
  • Each gage block features blended radii and is burr-proofed.
  • Measuring surfaces remain undamaged during wringing or stacking.
  • Includes a certificate of calibration traceable to NIST.
  • Ensures durability and precision due to its design, preventing damage during use.
  • Provides reliable and accurate measurements, supported by NIST traceable calibration.
Catalog No. 50-005-11443 Supplier MSC Supplier No. 03266640
This individual rectangular gage block, crafted from steel, offers precise measurements with a size of 0.121 inch and a Grade 0 accuracy. It is engineered for high durability and reliable performance in diverse measurement applications.
  • Each gage block features blended radii and is burr-proofed.
  • Measuring surfaces remain undamaged during wringing or stacking.
  • Includes a certificate of calibration traceable to NIST.
  • Ensures durability and precision due to its design, preventing damage during use.
  • Provides reliable and accurate measurements, supported by NIST traceable calibration.