Learn More
MSC SPI Gage Block Set, Grade 0 Steel, 0.0625-2 in, 9-piece
Catalog No. 5000514616
Encompass
This gage block set provides precise measurements for calibrating micrometers and other instruments. It includes nine steel blocks with a range of 0.0625 to 2 inches, an optical flat, and is furnished in a wooden case. Each block is individually serialized and includes NIST traceable certification.
  • Grade 0 accuracy
  • Range of 0.0625 to 2 inches
  • Includes nine blocks and one flat
  • Flatness of 0.000004 inches
  • Parallelism of 0.00002 inches
  • Constructed from steel
  • Individually serialized blocks
  • Certificate of calibration traceable to NIST
  • Furnished in a wooden case
Catalog No. 50-005-14616 Supplier MSC Supplier No. 69815330
This gage block set provides precise measurements for calibrating micrometers and other instruments. It includes nine steel blocks with a range of 0.0625 to 2 inches, an optical flat, and is furnished in a wooden case. Each block is individually serialized and includes NIST traceable certification.
  • Grade 0 accuracy
  • Range of 0.0625 to 2 inches
  • Includes nine blocks and one flat
  • Flatness of 0.000004 inches
  • Parallelism of 0.00002 inches
  • Constructed from steel
  • Individually serialized blocks
  • Certificate of calibration traceable to NIST
  • Furnished in a wooden case