Learn More
MSC SPI Gage Block Set, Rectangular, Grade 0, Steel, 5 to 20 in
Catalog No. 5000511681
Encompass
This gage block set provides precise measurements for calibration and inspection tasks. It features 8 steel, rectangular blocks with a Grade 0 accuracy, offering a measurement range from 5 to 20 inches.
  • 8 piece set
  • Rectangular shape
  • Steel material
  • Grade 0 accuracy
  • Individually serialized
  • NIST traceable calibration certificate included
  • Furnished in a fitted case
Catalog No. 50-005-11681 Supplier MSC Supplier No. 87877700
This gage block set provides precise measurements for calibration and inspection tasks. It features 8 steel, rectangular blocks with a Grade 0 accuracy, offering a measurement range from 5 to 20 inches.
  • 8 piece set
  • Rectangular shape
  • Steel material
  • Grade 0 accuracy
  • Individually serialized
  • NIST traceable calibration certificate included
  • Furnished in a fitted case