Learn More
MSC SPI Rectangle Steel Gage Block, Grade 0, 0.11 in., 1 ea.
Catalog No. 5000510967
Encompass
This rectangle steel gage block is designed for precision measurement applications and features a nominal size of 0.11000 in. Constructed from high-quality steel, it is manufactured to Grade 0 standards and includes a NIST-traceable certificate of calibration.
  • Accuracy grade 0
  • Steel construction
  • Includes NIST certification
  • Burr-proof design
  • Blended radii for improved performance
  • Wringing or stacking the block will not cause damage to the measuring surfaces
Catalog No. 50-005-10967 Supplier MSC Supplier No. 03266533
This rectangle steel gage block is designed for precision measurement applications and features a nominal size of 0.11000 in. Constructed from high-quality steel, it is manufactured to Grade 0 standards and includes a NIST-traceable certificate of calibration.
  • Accuracy grade 0
  • Steel construction
  • Includes NIST certification
  • Burr-proof design
  • Blended radii for improved performance
  • Wringing or stacking the block will not cause damage to the measuring surfaces