Learn More
MSC SPI Hardness Tester, Digital, 1-2967 HV, 0.1 HRC Resolution
Catalog No. 5000528217
Encompass
This bench top hardness tester is a digital instrument designed for precise hardness measurements. It features a broad hardness range from 1 HV to 2967 HV and offers a resolution of 0.1 HRC, making it suitable for various material testing applications.
  • Measures hardness on Knoop and Vickers scales.
  • Offers a wide hardness range from 1 HV to 2967 HV.
  • Equipped with a digital display for clear readings.
  • Provides high resolution of 0.1 HRC.
  • Compact design with 20.0 in depth and 10.0 in width.
Catalog No. 50-005-28217 Supplier MSC Supplier No. 38443339
This bench top hardness tester is a digital instrument designed for precise hardness measurements. It features a broad hardness range from 1 HV to 2967 HV and offers a resolution of 0.1 HRC, making it suitable for various material testing applications.
  • Measures hardness on Knoop and Vickers scales.
  • Offers a wide hardness range from 1 HV to 2967 HV.
  • Equipped with a digital display for clear readings.
  • Provides high resolution of 0.1 HRC.
  • Compact design with 20.0 in depth and 10.0 in width.