Learn More
MSC SPI Hardness Testing File, 50 RC, Rockwell C Scale, Light Green
Catalog No. 5000534365
Encompass
This hardness testing file provides a practical and inexpensive method for quickly determining the approximate hardness of heat-treated workpieces and materials. Designed for use with most bench-type Rockwell and Rockwell Superficial Type Hardness Testers, it is considered indispensable for machinists and heat treaters.
  • Special curved design for easy access in hard to reach places.
  • Quickly determine approximate hardness of heat-treated workpieces and materials.
  • 50 RC maximum hardness.
  • Light green color for easy identification.
Catalog No. 50-005-34365 Supplier MSC Supplier No. 00692541
This hardness testing file provides a practical and inexpensive method for quickly determining the approximate hardness of heat-treated workpieces and materials. Designed for use with most bench-type Rockwell and Rockwell Superficial Type Hardness Testers, it is considered indispensable for machinists and heat treaters.
  • Special curved design for easy access in hard to reach places.
  • Quickly determine approximate hardness of heat-treated workpieces and materials.
  • 50 RC maximum hardness.
  • Light green color for easy identification.